ENA313839192 Enamine

1-[5-fluoro-2-(1H-pyrazol-1-yl)phenyl]ethan-1-amine

  • MDL number MFCD11137162
  • Cas Number : 1152880-22-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 88
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-72729

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-72729
Availability details for
Quantity
  • Availability
  • Add to Cart