ENA313839199 Enamine

(2S)-2-hydroxy-2-phenylpropanoic acid

  • MDL number MFCD00067700
  • Cas Number : 13113-71-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Assay 95
Storage temp. 0

Safety Information

signalword Warning
msds https://www.enaminestore.com/catalog/EN300-72763
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart