ENA313839201 Enamine

6-hydrazinyl-1H,4H,5H-pyrazolo[3,4-d]pyrimidin-4-one

  • MDL number MFCD12966569
  • Cas Number : 98021-51-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-72773

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-72773
Availability details for
Quantity
  • Availability
  • Add to Cart