ENA313839210 Enamine

3-(2-methoxyphenyl)cyclobutan-1-one

  • MDL number MFCD11848524
  • Cas Number : 335331-55-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-43646

Safety Information

msds https://www.enaminestore.com/catalog/EN300-43646
Availability details for
Quantity
  • Availability
  • Add to Cart