ENA313839214 Enamine

3-(methoxycarbonyl)cyclohexane-1-carboxylic acid

  • MDL number MFCD17170191
  • Cas Number : 25090-39-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-43933

Safety Information

msds https://www.enaminestore.com/catalog/EN300-43933
Availability details for
Quantity
  • Availability
  • Add to Cart