ENA313839221 Enamine

N'-hydroxy-2-(thiophen-2-yl)ethanimidamide

  • Cas Number : 59174-12-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 79-82
Purity 95
Product Link https://www.enaminestore.com/catalog/EN300-72806

Safety Information

signalword Warning
pcodes P261,P264,P271,P280,P302+P352,P304+P340,P305+P351+P338,P312,P332+P313,P337+P313,P362,P403+P233,P405,P501
hcodes Xi
MSDS https://www.enaminestore.com/catalog/EN300-72806
pictograms GHS07
hstatements H315,H319,H335
Availability details for
Quantity
  • Availability
  • Add to Cart