ENA313839226 Enamine

4-chloro-1-(prop-2-yn-1-yl)-1H-pyrazole

  • MDL number MFCD14638461
  • Cas Number : 1251029-14-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-72812

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-72812
Availability details for
Quantity
  • Availability
  • Add to Cart