ENA313839227 Enamine

1-(isoquinolin-5-yl)-2,5-dihydro-1H-pyrrole-2,5-dione

  • MDL number MFCD12066293
  • Cas Number : 1157377-28-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-44329

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-44329
Availability details for
Quantity
  • Availability
  • Add to Cart