ENA313839235 Enamine

tert-butyl N-(1H-pyrazol-4-yl)carbamate

  • MDL number MFCD12133944
  • Cas Number : 130106-42-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 179...181
Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-64880

Safety Information

signalword Warning
msds https://www.enaminestore.com/catalog/EN300-64880
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart