ENA313839240 Enamine

4-[(2-methanesulfonylethane)sulfonyl]aniline

  • MDL number MFCD14643844
  • Cas Number : 98960-57-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 209...211
Purity 95.00
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-72833

Safety Information

msds https://www.enaminestore.com/catalog/EN300-72833
Availability details for
Quantity
  • Availability
  • Add to Cart