ENA313839243 Enamine

2-(2,4-dichlorophenyl)-1-(4-fluorophenyl)ethan-1-amine

  • MDL number MFCD12605454
  • Cas Number : 1179226-43-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-72838

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-72838
Availability details for
Quantity
  • Availability
  • Add to Cart