ENA313839248 Enamine

8-tert-butyl-2-azaspiro[4.5]decane-1,3-dione

  • MDL number MFCD03237428
  • Cas Number : 183120-06-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-66684

Safety Information

msds https://www.enaminestore.com/catalog/EN300-66684
Availability details for
Quantity
  • Availability
  • Add to Cart