ENA313839249 Enamine

2-chloro-N-(cyclobutylmethyl)acetamide

  • MDL number MFCD11634955
  • Cas Number : 1095592-81-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-72861

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-72861
Availability details for
Quantity
  • Availability
  • Add to Cart