ENA313839262 Enamine

4-methyl-6-(propan-2-yl)-1,3,5-triazin-2-amine

  • MDL number MFCD18380673
  • Cas Number : 82020-89-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-72888

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-72888
Availability details for
Quantity
  • Availability
  • Add to Cart