ENA313839266 Enamine

N1-cyclopentylbenzene-1,3-diamine

  • MDL number MFCD11101551
  • Cas Number : 506436-21-1
  • Molecular Weight : 176.26

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form oil
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-72892

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-72892
Availability details for
Quantity
  • Availability
  • Add to Cart