ENA313839268 Enamine

N-{[2-(methoxymethyl)phenyl]methyl}-1,3-benzoxazol-2-amine

  • MDL number MFCD11830202
  • Cas Number : 1090598-92-7
  • Molecular Weight : 268.32

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-72897

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-72897
Availability details for
Quantity
  • Availability
  • Add to Cart