ENA313839277 Enamine

2-phenylmethanesulfinylethan-1-amine

  • MDL number MFCD14612871
  • Cas Number : 30491-32-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-72909

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-72909
Availability details for
Quantity
  • Availability
  • Add to Cart