ENA313839286 Enamine

5-amino-2-(2,2,2-trifluoroethoxy)benzoic acid

  • MDL number MFCD12168743
  • Cas Number : 1096332-51-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Product Link https://www.enaminestore.com/catalog/EN300-72919

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-72919
Availability details for
Quantity
  • Availability
  • Add to Cart