ENA313839291 Enamine

2-methyl-2-phenylpropanehydrazide

  • MDL number MFCD09802904
  • Cas Number : 5809-15-4
  • Molecular Weight : 178.23

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-72937

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-72937
Availability details for
Quantity
  • Availability
  • Add to Cart