ENA313839292 Enamine

5-(dimethylamino)-1,3,4-thiadiazole-2-thiol

  • MDL number MFCD16103265
  • Cas Number : 2810-66-4
  • Molecular Weight : 161.25

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-72938

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-72938
Availability details for
Quantity
  • Availability
  • Add to Cart