ENA313839299 Enamine

2-amino-3-methylbutanenitrile hydrochloride

  • MDL number MFCD18380682
  • Cas Number : 60586-49-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-72947

Safety Information

msds https://www.enaminestore.com/catalog/EN300-72947
Availability details for
Quantity
  • Availability
  • Add to Cart