ENA313839305 Enamine

tert-butyl 3-(cyclobutylamino)piperidine-1-carboxylate

  • MDL number MFCD06656624
  • Cas Number : 887586-09-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-72955

Safety Information

msds https://www.enaminestore.com/catalog/EN300-72955
Availability details for
Quantity
  • Availability
  • Add to Cart