ENA313839309 Enamine

1-(4-bromo-2-fluorophenyl)propan-2-amine

  • MDL number MFCD08452923
  • Cas Number : 910409-78-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Product Link https://www.enaminestore.com/catalog/EN300-72959

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-72959
Availability details for
Quantity
  • Availability
  • Add to Cart