ENA313839310 Enamine

3H,4H,5H,6H,7H,8H-pyrido[2,3-d]pyrimidin-4-one

  • MDL number MFCD18380603
  • Cas Number : 3649-42-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 243-245
Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-71364

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-71364
Availability details for
Quantity
  • Availability
  • Add to Cart