ENA313839322 Enamine

3-(3-bromophenyl)propan-1-amine hydrochloride

  • MDL number MFCD09260975
  • Cas Number : 1269151-20-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-73002

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-73002
Availability details for
Quantity
  • Availability
  • Add to Cart