ENA313839325 Enamine

3-(5-phenyl-2H-1,2,3,4-tetrazol-2-yl)propan-1-ol

  • MDL number MFCD12179149
  • Cas Number : 1153022-33-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Product Link https://www.enaminestore.com/catalog/EN300-73006

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-73006
Availability details for
Quantity
  • Availability
  • Add to Cart