ENA313839328 Enamine

1-acetamidocyclopentane-1-carboxylic acid

  • MDL number MFCD01735322
  • Cas Number : 4854-46-0
  • Molecular Weight : 171.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-73011

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-73011
Availability details for
Quantity
  • Availability
  • Add to Cart