ENA313839331 Enamine

4-chloro-2-(pyridin-3-yl)-6-(trifluoromethyl)pyrimidine

  • MDL number MFCD00140057
  • Cas Number : 204394-69-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-71873

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-71873
Availability details for
Quantity
  • Availability
  • Add to Cart