ENA313839336 Enamine

1-[4-(phenylamino)piperidin-1-yl]ethan-1-one

  • MDL number MFCD12049473
  • Cas Number : 1157294-82-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Product Link https://www.enaminestore.com/catalog/EN300-73027

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-73027
Availability details for
Quantity
  • Availability
  • Add to Cart