ENA313839338 Enamine

3-(2-chlorophenyl)-1-methyl-1H-pyrazole-4-carboxylic acid

  • MDL number MFCD11103111
  • Cas Number : 934405-77-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-73030

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-73030
Availability details for
Quantity
  • Availability
  • Add to Cart