ENA313839339 Enamine

1-tert-butyl-3-(2-chlorophenyl)-1H-pyrazole-4-carbaldehyde

  • MDL number MFCD11539834
  • Cas Number : 1152874-49-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Product Link https://www.enaminestore.com/catalog/EN300-73032

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-73032
Availability details for
Quantity
  • Availability
  • Add to Cart