ENA313839341 Enamine

2-(1H-1,2,3,4-tetrazol-5-yl)pyridine

  • MDL number MFCD00068114
  • Cas Number : 33893-89-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 210-212
Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-73036

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-73036
Availability details for
Quantity
  • Availability
  • Add to Cart