ENA313839353 Enamine

3-(3-hydroxypropyl)-1,3-diazaspiro[4.5]decane-2,4-dione

  • MDL number MFCD12186696
  • Cas Number : 1153129-25-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Product Link https://www.enaminestore.com/catalog/EN300-73049

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-73049
Availability details for
Quantity
  • Availability
  • Add to Cart