ENA313839354 Enamine

1-(3-hydroxyphenyl)-2,5-dihydro-1H-pyrrole-2,5-dione

  • MDL number MFCD00461770
  • Cas Number : 63381-38-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-73050

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-73050
Availability details for
Quantity
  • Availability
  • Add to Cart