ENA313839355 Enamine

2-[(piperazine-1-sulfonyl)amino]ethan-1-ol

  • MDL number MFCD14605408
  • Cas Number : 1247805-31-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-73051

Safety Information

msds https://www.enaminestore.com/catalog/EN300-73051
Availability details for
Quantity
  • Availability
  • Add to Cart