ENA313839359 Enamine

5-methyl-1-(quinolin-8-yl)-1H-1,2,3-triazole-4-carboxylic acid

  • MDL number MFCD12739197
  • Cas Number : 1094246-02-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 188...190
Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-73058

Safety Information

msds https://www.enaminestore.com/catalog/EN300-73058
Availability details for
Quantity
  • Availability
  • Add to Cart