ENA313839361 Enamine

2-amino-5-chloro-N-ethylbenzamide

  • MDL number MFCD09048143
  • Cas Number : 32212-30-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-73060

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-73060
Availability details for
Quantity
  • Availability
  • Add to Cart