ENA313839362 Enamine

1-methyl-1-[(thiophen-2-yl)methyl]urea

  • MDL number MFCD12189990
  • Cas Number : 1153872-31-1
  • Molecular Weight : 170.24

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-73061

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-73061
Availability details for
Quantity
  • Availability
  • Add to Cart