ENA313839371 Enamine

1-(2,4-dichlorophenyl)butan-2-amine

  • MDL number MFCD11937066
  • Cas Number : 1004284-08-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-73072

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-73072
Availability details for
Quantity
  • Availability
  • Add to Cart