ENA313839390 Enamine

2-(2-methylpropoxy)benzoic acid

  • MDL number MFCD06800806
  • Cas Number : 147578-43-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-73106

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-73106
Availability details for
Quantity
  • Availability
  • Add to Cart