ENA313839391 Enamine

N-(3-aminophenyl)-3-(piperidin-1-yl)propanamide

  • MDL number MFCD09729632
  • Cas Number : 858262-64-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-73107

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-73107
Availability details for
Quantity
  • Availability
  • Add to Cart