ENA313839397 Enamine

3-amino-4-methanesulfonylbenzonitrile

  • MDL number MFCD12189552
  • Cas Number : 1153524-12-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 150...152
Purity 95.00
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-72113

Safety Information

msds https://www.enaminestore.com/catalog/EN300-72113
Availability details for
Quantity
  • Availability
  • Add to Cart