ENA313839403 Enamine

N1-(4-methyl-1,3-thiazol-2-yl)benzene-1,4-diamine

  • MDL number MFCD09941772
  • Cas Number : 1745-19-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-72218

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-72218
Availability details for
Quantity
  • Availability
  • Add to Cart