ENA313839405 Enamine

2-tert-butyl-6-methylpyrimidin-4-ol

  • MDL number MFCD09881380
  • Cas Number : 90565-53-0
  • Molecular Weight : 166.22

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-73124

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-73124
Availability details for
Quantity
  • Availability
  • Add to Cart