ENA313839407 Enamine

2-(2-methylphenyl)-3-oxo-5-phenylpentanenitrile

  • MDL number MFCD13658643
  • Cas Number : 1097048-05-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-73127

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-73127
Availability details for
Quantity
  • Availability
  • Add to Cart