ENA313839409 Enamine

methyl 4-methanesulfonylbutanoate

  • MDL number MFCD12823563
  • Cas Number : 67399-14-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-73129

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-73129
Availability details for
Quantity
  • Availability
  • Add to Cart