ENA313839418 Enamine

3,6-dichloropyridazine-4-carboxamide

  • MDL number MFCD11646064
  • Cas Number : 27427-66-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 175-179
Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-73145

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-73145
Availability details for
Quantity
  • Availability
  • Add to Cart