ENA313839421 Enamine

2,4,4-trimethyl-3-oxopentanenitrile

  • MDL number MFCD11105088
  • Cas Number : 29509-04-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-73150

Safety Information

msds https://www.enaminestore.com/catalog/EN300-73150
Availability details for
Quantity
  • Availability
  • Add to Cart