ENA313839433 Enamine

2-(piperazin-1-yl)-1-(1,2,3,4-tetrahydroquinolin-1-yl)ethan-1-one

  • MDL number MFCD10011127
  • Cas Number : 1018568-58-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-73168

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-73168
Availability details for
Quantity
  • Availability
  • Add to Cart