ENA313839437 Enamine

N-(1H-pyrazol-3-yl)benzamide

  • MDL number MFCD14657344
  • Cas Number : 54135-43-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Product Link https://www.enaminestore.com/catalog/EN300-73174

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-73174
Availability details for
Quantity
  • Availability
  • Add to Cart